Learn More
Med Vet International Dynarex Safe-Touch Latex Exam Gloves, Small, Powder-Free, 1000/Case
Catalog No. 502830041
Encompass_Preferred
Dynarex Safe-Touch Latex Exam Gloves are premium-quality, powder-free latex gloves crafted for optimal protection and comfort. The ambidextrous Gloves are designed with textured fingertips for increased grip and a 3 mil minimum thickness for improved tactile sensitivity. The Exam Gloves feature a polymer lining for added durability and beaded cuffs to ensure easy donning and help resist tears. These Gloves have a secure fit around the wrist and hand-conforming flexibility to help prevent physical fatigue. Ideal for many settings including veterinary clinics, retirement homes, and medical facilities. Dynarex Safe-Touch Latex Exam Gloves are high-quality, polymer-lined gloves designed for enhanced protection and performance. These ambidextrous, powder-free Gloves feature a 3 mil minimum thickness for improved sensitivity, textured fingertips for increased grip, and beaded cuffs to allow for easy donning. Ideal for a wide range of healthcare and professional environments.
Catalog No. 50-283-0041 Supplier Med Vet International Supplier No. 2336
Dynarex Safe-Touch Latex Exam Gloves are premium-quality, powder-free latex gloves crafted for optimal protection and comfort. The ambidextrous Gloves are designed with textured fingertips for increased grip and a 3 mil minimum thickness for improved tactile sensitivity. The Exam Gloves feature a polymer lining for added durability and beaded cuffs to ensure easy donning and help resist tears. These Gloves have a secure fit around the wrist and hand-conforming flexibility to help prevent physical fatigue. Ideal for many settings including veterinary clinics, retirement homes, and medical facilities. Dynarex Safe-Touch Latex Exam Gloves are high-quality, polymer-lined gloves designed for enhanced protection and performance. These ambidextrous, powder-free Gloves feature a 3 mil minimum thickness for improved sensitivity, textured fingertips for increased grip, and beaded cuffs to allow for easy donning. Ideal for a wide range of healthcare and professional environments.