Learn More
Med Vet International Dynarex Sensi Grip Latex Exam Gloves, Small, Powder-Free, 1000/Case
Supplier:  Med Vet International 6712
Encompass_Preferred
Dynarex Sensi Grip Latex Exam Gloves are premium-quality, powder-free latex gloves crafted for optimal protection and comfort. The ambidextrous Gloves are designed with textured fingertips for increased grip and 3 mil minimum thickness for enhanced tactile sensitivity. The Gloves are chlorinated to help provide a smooth, easy fit. A polymer lining provides improved tear-resistance and beaded cuffs allow for easy donning. Ideal for a wide variety of settings, including law enforcement, correctional facilities, tattoo shops, hair salons, first responders, and medical facilities.  Dynarex Sensi Grip Latex Exam Gloves are chlorinated, polymer-lined gloves designed for enhanced protection and performance. These ambidextrous, powder-free Gloves feature a 3 mil minimum thickness for tactile sensitivity, textured fingertips for increased grip, and beaded cuffs to allow for easy donning. 
Catalog No. 50-283-1901