Learn More
Med Vet International Dynarex Sterile Latex Exam Gloves, Large, Powder-Free, 400/Case
Catalog No. 502830620
Encompass_Preferred
Dynarex Sterile Latex Exam Gloves are premium-quality, powder-free latex gloves crafted for optimal protection and performance. The Gloves are designed with textured palms for increased grip and 3 mil minimum thickness for increased tactile sensitivity. The Exam Gloves feature beaded cuffs to help ensure easy donning. These Gloves have a secure fit around the wrist and hand-conforming flexibility to help prevent physical fatigue. Dynarex Sterile Latex Exam Gloves come in pairs and are sterilized with Ethylene Oxide (EO). Ideal for a variety of settings, including medical facilities, veterinary clinics, and long-term care facilities. Dynarex Sterile Latex Exam Gloves are high-quality sterile gloves designed for enhanced comfort and protection. These powder-free Gloves feature a 3 mil minimum thickness to allow for tactile sensitivity, textured palms for increased grip, and beaded cuffs to allow for easy donning. 
Catalog No. 50-283-0620 Supplier Med Vet International Supplier No. 2453
Dynarex Sterile Latex Exam Gloves are premium-quality, powder-free latex gloves crafted for optimal protection and performance. The Gloves are designed with textured palms for increased grip and 3 mil minimum thickness for increased tactile sensitivity. The Exam Gloves feature beaded cuffs to help ensure easy donning. These Gloves have a secure fit around the wrist and hand-conforming flexibility to help prevent physical fatigue. Dynarex Sterile Latex Exam Gloves come in pairs and are sterilized with Ethylene Oxide (EO). Ideal for a variety of settings, including medical facilities, veterinary clinics, and long-term care facilities. Dynarex Sterile Latex Exam Gloves are high-quality sterile gloves designed for enhanced comfort and protection. These powder-free Gloves feature a 3 mil minimum thickness to allow for tactile sensitivity, textured palms for increased grip, and beaded cuffs to allow for easy donning. 