Learn More
Med Vet International Dynarex Accutouch Latex Exam Gloves, X-Small, Powder-Free, 1000/Case
Supplier:  Med Vet International 6621
Encompass_Preferred
Dynarex AccuTouch Latex Exam Gloves are premium-quality, powder-free latex gloves crafted for optimal protection and comfort. The ambidextrous Gloves are designed with textured palms for increased grip and a minimum thickness of 4 mil at the palm and 5 mil at the fingertips for enhanced tactile sensitivity. The Gloves feature a polymer lining for improved tear-resistance and beaded cuffs to help ensure easy donning. These Exam Gloves have a secure fit around the wrist and h and -conforming flexibility to help prevent physical fatigue. Ideal for a wide variety of settings, including law enforcement, correctional facilities, and medical facilities. Dynarex AccuTouch Latex Exam Gloves are high-quality, polymer-lined gloves designed for enhanced protection and performance. These ambidextrous, powder-free Gloves feature a minimum thickness of 4 mil at the palm and 5 mil at the fingertips for tactile sensitivity, textured palms for increased grip, and beaded cuffs to allow for easy donning.
Catalog No. 50-283-0021