Learn More
Electron Microscopy Sciences Latex Gloves Powder Free, Medium
SDP
Catalog No. 501899495
Encompass_Preferred
Made from 100% natural latex with the addition of Aloe Vera and Vitamin E, offering the softness, tactile sensitivity and skin protection. Economical ambidextrous style, beaded cuff, and are chemically resistant to acids, alcohols, ketones and alkalis. Packed in a convenient dispenser box. Non-sterile. Non-sterile, powder free.
Size: Medium
This item is not returnable. View return policy
Catalog No. 50-189-9495 Supplier Electron Microscopy Sciences Supplier No. 7111501
Made from 100% natural latex with the addition of Aloe Vera and Vitamin E, offering the softness, tactile sensitivity and skin protection. Economical ambidextrous style, beaded cuff, and are chemically resistant to acids, alcohols, ketones and alkalis. Packed in a convenient dispenser box. Non-sterile. Non-sterile, powder free.
Size: Medium