Learn More
Cole-Parmer PIP 730-M Glove liners, full-fingered, medium, 12 pairs/pk
Catalog No. NC0429813 Shop All Cole-Parmer Products
Encompass_Preferred
Experience the softness and comfort of seamless, fine-gauge nylon knit. Dust-free nylon is ideal for cleanrooms. Easily wash and sterilize for repeated use.
This item is not returnable. View return policy
Catalog No. NC0429813 Supplier Cole-Parmer Supplier No. 8638201
Experience the softness and comfort of seamless, fine-gauge nylon knit. Dust-free nylon is ideal for cleanrooms. Easily wash and sterilize for repeated use.