Learn More
Med Vet International Dynarex Safe-Touch Nitrile Exam Gloves, Large, Powder-Free, Black, 1000/Case
Catalog No. 502831415
Encompass_Preferred
Dynarex Safe-Touch Black Nitrile Exam Gloves are premium-quality powder-free nitrile gloves crafted for optimal protection and tactile sensitivity. The ambidextrous Gloves are designed with textured fingertips for increased grip and feature a thickness range of 3-4 mils at the palms and 4-6 mils at the fingertips. The Exam Gloves feature a polymer lining for improved puncture-resistance and beaded cuff to ensure easy donning. These Gloves have a secure fit around the wrist and h and -conforming flexibility to help prevent physical fatigue. Ideal for a wide variety of settings including medical facilities EMS and beauty salons. Dynarex Safe-Touch Black Nitrile Exam Gloves are high-quality polymer-lined gloves designed for enhanced protection and comfort. These ambidextrous powder-free gloves feature a thickness range of 3-4 mils at the palms and 4-6 mils at the fingertips. The gloves feature textured fingertips for increased grip and beaded cuffs to allow for easy donning.  
Catalog No. 50-283-1415 Supplier Med Vet International Supplier No. DYN2523
Dynarex Safe-Touch Black Nitrile Exam Gloves are premium-quality powder-free nitrile gloves crafted for optimal protection and tactile sensitivity. The ambidextrous Gloves are designed with textured fingertips for increased grip and feature a thickness range of 3-4 mils at the palms and 4-6 mils at the fingertips. The Exam Gloves feature a polymer lining for improved puncture-resistance and beaded cuff to ensure easy donning. These Gloves have a secure fit around the wrist and h and -conforming flexibility to help prevent physical fatigue. Ideal for a wide variety of settings including medical facilities EMS and beauty salons. Dynarex Safe-Touch Black Nitrile Exam Gloves are high-quality polymer-lined gloves designed for enhanced protection and comfort. These ambidextrous powder-free gloves feature a thickness range of 3-4 mils at the palms and 4-6 mils at the fingertips. The gloves feature textured fingertips for increased grip and beaded cuffs to allow for easy donning.  