Learn More
Med Vet International Dynarex Aloeskin Nitrile Exam Gloves w/Aloe, Large, Rose, Powder-Free, 1000/Case
Catalog No. 502829957
Encompass_Preferred
Dynarex AloeSkin Nitrile Exam Gloves are premium-quality, powder-free nitrile gloves crafted for optimal protection and comfort. To help nourish skin, these Gloves are infused with soothing aloe and vitamin E. The ambidextrous magenta Gloves are designed with textured fingertips for increased grip and feature a 2 mil minimum thickness at fingertips and palms for increased tactile sensitivity. These Exam Gloves feature a polymer lining for improved puncture-resistance and beaded cuffs to ensure easy donning. The Gloves have a secure fit around the wrist and hand-conforming flexibility to help prevent physical fatigue. Ideal for a wide range of healthcare facilities and professional environments. Dynarex AloeSkin Nitrile Exam Gloves are powder-free, polymer-lined gloves designed for enhanced protection and performance. These ambidextrous Gloves are coated with skin-soothing aloe and vitamin E and feature a minimum thickness of 2 mil.
Catalog No. 50-282-9957 Supplier Med Vet International Supplier No. 6723
Dynarex AloeSkin Nitrile Exam Gloves are premium-quality, powder-free nitrile gloves crafted for optimal protection and comfort. To help nourish skin, these Gloves are infused with soothing aloe and vitamin E. The ambidextrous magenta Gloves are designed with textured fingertips for increased grip and feature a 2 mil minimum thickness at fingertips and palms for increased tactile sensitivity. These Exam Gloves feature a polymer lining for improved puncture-resistance and beaded cuffs to ensure easy donning. The Gloves have a secure fit around the wrist and hand-conforming flexibility to help prevent physical fatigue. Ideal for a wide range of healthcare facilities and professional environments. Dynarex AloeSkin Nitrile Exam Gloves are powder-free, polymer-lined gloves designed for enhanced protection and performance. These ambidextrous Gloves are coated with skin-soothing aloe and vitamin E and feature a minimum thickness of 2 mil.