Learn More
Med Vet International Dynarex Dynaplus Nitrile Exam Gloves, Large, Powder-Free, Violet, 2000/Case
Supplier:  Med Vet International 2518
Encompass_Preferred
Dynarex DynaPlus Nitrile Exam Gloves are premium-quality, powder-free nitrile gloves crafted for optimal protection and comfort. The ambidextrous Gloves are designed with textured fingertips for increased grip and a minimum thickness of 2 mil at the palm and fingertips for increased tactile sensitivity. The Exam Gloves feature beaded cuffs to help ensure easy donning. These Gloves have a secure fit around the wrist and hand-conforming flexibility to help prevent physical fatigue. Ideal for use in a wide variety of settings and industries, including medical facilities, EMS, law enforcement, long-term care, tattoo studios, and beauty salons.  Dynarex DynaPlus Nitrile Exam Gloves are high-quality nitrile gloves designed for enhanced protection and performance. These ambidextrous, powder-free Gloves feature a minimum thickness of 2 mil at the palm and fingertips for tactile sensitivity, textured fingertips for increased grip, and beaded cuffs to allow for easy donning. 
Catalog No. 50-283-0111