Learn More
Med Vet International Dynarex Safe-Touch Nitrile Exam Gloves, Medium, Powder-Free, Black, 1000/Case
Catalog No. 502830114
Encompass_Preferred
Dynarex Safe-Touch Black Nitrile Exam Gloves are premium-quality, powder-free nitrile gloves crafted for optimal protection and tactile sensitivity. The ambidextrous Gloves are designed with textured fingertips for increased grip and feature a thickness range of 3-4 mils at the palms and 4-6 mils at the fingertips. These Gloves have a secure fit around the wrist and hand-conforming flexibility to help prevent physical fatigue. Ideal for a wide variety of settings, including medical facilities, EMS, law enforcement, long-term care, tattoo studios, and beauty salons. Dynarex Safe-Touch Black Nitrile Exam Gloves are high-quality, polymer-lined gloves designed for enhanced protection and comfort. These ambidextrous, powder-free gloves feature a thickness range of 3-4 mils at the palms and 4-6 mils at the fingertips. The gloves feature textured fingertips for increased grip and beaded cuffs to allow for easy donning. 
Catalog No. 50-283-0114 Supplier Med Vet International Supplier No. 2522
Dynarex Safe-Touch Black Nitrile Exam Gloves are premium-quality, powder-free nitrile gloves crafted for optimal protection and tactile sensitivity. The ambidextrous Gloves are designed with textured fingertips for increased grip and feature a thickness range of 3-4 mils at the palms and 4-6 mils at the fingertips. These Gloves have a secure fit around the wrist and hand-conforming flexibility to help prevent physical fatigue. Ideal for a wide variety of settings, including medical facilities, EMS, law enforcement, long-term care, tattoo studios, and beauty salons. Dynarex Safe-Touch Black Nitrile Exam Gloves are high-quality, polymer-lined gloves designed for enhanced protection and comfort. These ambidextrous, powder-free gloves feature a thickness range of 3-4 mils at the palms and 4-6 mils at the fingertips. The gloves feature textured fingertips for increased grip and beaded cuffs to allow for easy donning. 