Learn More
Med Vet International Dynarex True Advantage Nitrile Exam Gloves, Medium, Powder-Free, 1000/Case
Catalog No. 502830120
Encompass_Preferred
Dynarex True Advantage Nitrile Exam Gloves are chemotherapy approved, powder-free nitrile gloves crafted for optimal protection and puncture resistance. The premium-quality ambidextrous Gloves feature 3-mil minimum thickness at fingertips and palms for increased tactile sensitivity. The Exam Gloves feature beaded cuffs to ensure easy donning. These Gloves have a secure fit around the wrist and hand-conforming flexibility to help prevent physical fatigue. Ideal for a wide variety of settings, including long-term care facilities, hospitals, emergency situations, and more.   Dynarex True Advantage Nitrile Exam Gloves are premium-quality gloves designed for enhanced protection and comfort. These ambidextrous, powder-free gloves feature beaded cuffs and a minimum thickness of 3 mil. 
Catalog No. 50-283-0120 Supplier Med Vet International Supplier No. 6503
Dynarex True Advantage Nitrile Exam Gloves are chemotherapy approved, powder-free nitrile gloves crafted for optimal protection and puncture resistance. The premium-quality ambidextrous Gloves feature 3-mil minimum thickness at fingertips and palms for increased tactile sensitivity. The Exam Gloves feature beaded cuffs to ensure easy donning. These Gloves have a secure fit around the wrist and hand-conforming flexibility to help prevent physical fatigue. Ideal for a wide variety of settings, including long-term care facilities, hospitals, emergency situations, and more.   Dynarex True Advantage Nitrile Exam Gloves are premium-quality gloves designed for enhanced protection and comfort. These ambidextrous, powder-free gloves feature beaded cuffs and a minimum thickness of 3 mil. 