Learn More
Med Vet International Dynarex Safe-Touch Blue Nitrile Exam Gloves - Non-Latex, Small, Powder-Free, Blue, 1000/Case
Supplier:  Med Vet International 2511
Encompass_Preferred
Dynarex Safe-Touch Blue Nitrile Exam Gloves are premium-quality, powder-free nitrile gloves crafted for optimal protection and comfort. These ambidextrous Gloves are designed with textured fingertips for improved grip. The Nitrile Exam Gloves feature a thickness range of 3-4 mil at the palm and 4-6 mil at fingertips. The Exam Gloves feature a polymer lining and beaded cuff to help ensure easy donning and help prevent tears. These Gloves have a secure fit around the wrist and h and -conforming flexibility to help prevent physical fatigue. Ideal for a wide variety of settings, including medical facilities, EMS and law enforcement, long-term care, tattoo shops, hair salons, and more. Dynarex Safe-Touch Blue Nitrile Exam Gloves are high-quality, polymer-lined gloves designed for enhanced protection and performance. The Nitrile Exam Gloves have a thickness range of 3-4 mil at the palm and 4-6 mil at fingertips to provide optimal tactile sensitivity and puncture resistance.
Catalog No. 50-283-0621