Learn More
Med Vet International Dynarex Nitrile Sterile Exam Gloves, Singles, Small, Powder-Free, 800/Case
Supplier:  Med Vet International DYN6522
Encompass_Preferred
Dynarex Sterile Nitrile Exam Gloves are premium-quality, powder-free nitrile gloves crafted for optimal protection and performance. The ambidextrous Gloves are designed with textured palms for improved grip and beaded cuffs for easy donning. The Exam Gloves feature a 2 mil minimum thickness for increased tactile sensitivity. These nitrile Gloves have a secure fit around the wrist and hand-conforming flexibility to help prevent physical fatigue. These Sterile Gloves are packaged in individual peel-open packs for optimal hygiene. Ideal for a wide variety of settings, including medical facilities, EMS and law enforcement markets, long-term care facilities, and more. Dynarex Sterile Nitrile Exam Gloves are high-quality nitrile gloves designed for enhanced protection and comfort. These ambidextrous, powder-free gloves feature a minimum thickness of 2 mil for tactile sensitivity, textured palms for increased grip, and beaded cuffs to allow for easy donning.
Catalog No. 50-283-2012