Learn More
Med Vet International Dynarex Safe-Touch Nitrile Exam Gloves, X-Large, Powder-Free, Black, 1000/Case
Catalog No. 502832121
Encompass_Preferred
Dynarex Safe-Touch Black Nitrile Exam Gloves are premium-quality, powder-free nitrile gloves crafted for optimal protection and tactile sensitivity. The ambidextrous Gloves are designed with textured fingertips for increased grip and feature a thickness range of 3-4 mils at the palms and 4-6 mils at the fingertips. The Exam Gloves feature a polymer lining for improved puncture-resistance and beaded cuff to ensure easy donning. These Gloves have a secure fit around the wrist and h and -conforming flexibility to help prevent physical fatigue. Ideal for a wide variety of settings, including medical facilities, EMS, and beauty salons. Dynarex Safe-Touch Black Nitrile Exam Gloves are high-quality, polymer-lined gloves designed for enhanced protection and comfort. These ambidextrous, powder-free gloves feature a thickness range of 3-4 mils at the palms and 4-6 mils at the fingertips. The gloves feature textured fingertips for increased grip and beaded cuffs to allow for easy donning.  
Catalog No. 50-283-2121 Supplier Med Vet International Supplier No. 2524
Dynarex Safe-Touch Black Nitrile Exam Gloves are premium-quality, powder-free nitrile gloves crafted for optimal protection and tactile sensitivity. The ambidextrous Gloves are designed with textured fingertips for increased grip and feature a thickness range of 3-4 mils at the palms and 4-6 mils at the fingertips. The Exam Gloves feature a polymer lining for improved puncture-resistance and beaded cuff to ensure easy donning. These Gloves have a secure fit around the wrist and h and -conforming flexibility to help prevent physical fatigue. Ideal for a wide variety of settings, including medical facilities, EMS, and beauty salons. Dynarex Safe-Touch Black Nitrile Exam Gloves are high-quality, polymer-lined gloves designed for enhanced protection and comfort. These ambidextrous, powder-free gloves feature a thickness range of 3-4 mils at the palms and 4-6 mils at the fingertips. The gloves feature textured fingertips for increased grip and beaded cuffs to allow for easy donning.  