Learn More
Med Vet International Dynarex Nitrile Exam Gloves in A Bag, X-Large, Powder-Free, Blue, 2 Pairs/Bag, 500 Bags/Case
Supplier:  Med Vet International DYN2619
Encompass_Preferred
Dynarex DynaSafety Nitrile Gloves in a Bag are premium-quality, powder-free nitrile gloves crafted for optimal protection and comfort. The ambidextrous Gloves are designed with textured fingertips for increased grip and a minimum thickness of 2 mils at fingertips and palm for increased tactile sensitivity. The Gloves feature beaded cuffs to help ensure easy donning. These Gloves in a Bag have a secure fit around the wrist and hand-conforming flexibility to help prevent physical fatigue. The Nitrile Gloves are packaged in single or double pair bags. Ideal for use in a wide variety of settings, including hospitals, emergency situations, long-term care, tattoo studios, and veterinary practices. Dynarex DynaSafety Nitrile Gloves in a Bag are high-quality nitrile gloves designed for enhanced protection and tactile sensitivity. These ambidextrous, powder-free Gloves feature textured fingertips for increased grip and beaded cuffs to allow for easy donning.
Catalog No. 50-283-1507
 Restricted Item