Learn More
Med Vet International SW PowerForm PF-95BL Blue LargePowder Free Disposable Gloves - Medical Exam Grade - 9.5 in Length - Textured Fingers Finish - 5.0 mil Thick - N700344, 100/Box
Catalog No. 502522228
Encompass_Preferred
Product SpecificationsBrand: SW SafetyProduct Type: Disposable GlovesGrade: Medical ExamMaterial: NitrilePowdering: Powder FreeThickness: 5.0 milOverall Length: 9.5 inColor: BlueSize: LargeSurface Finish: Textured FingersResistant Properties: Chemical-ResistantCuff Type: BeadedSeries: PF-95BLStandards Met: SHA Dermatologically ApprovedPackage Quantity: 100 per box, 10 boxes per caseTrade Name: PowerForm, Dry Core• Dry Core absorbent technology keeps hands dry and cool• Fully textured surface delivers added grip• Beaded cuff allows easy donning and doffing• 100% air inspected / consistent thickness tip to cuff• ANSI Level 3 abrasion resistance
Catalog No. 50-252-2228 Supplier Med Vet International Supplier No. N700344
Product SpecificationsBrand: SW SafetyProduct Type: Disposable GlovesGrade: Medical ExamMaterial: NitrilePowdering: Powder FreeThickness: 5.0 milOverall Length: 9.5 inColor: BlueSize: LargeSurface Finish: Textured FingersResistant Properties: Chemical-ResistantCuff Type: BeadedSeries: PF-95BLStandards Met: SHA Dermatologically ApprovedPackage Quantity: 100 per box, 10 boxes per caseTrade Name: PowerForm, Dry Core• Dry Core absorbent technology keeps hands dry and cool• Fully textured surface delivers added grip• Beaded cuff allows easy donning and doffing• 100% air inspected / consistent thickness tip to cuff• ANSI Level 3 abrasion resistance