Learn More
Med Vet International Dynarex Nitrile Sterile Surgical Gloves, Pairs, Size 6.5, 200/Case
Supplier:  Med Vet International 6534
Encompass_Preferred
Dynarex Sterile Nitrile Surgical Gloves are premium-quality, powder-free nitrile gloves crafted for optimal protection and comfort. These hand-specific Gloves feature a thickness range of 5-8 mil at the palm and fingertips. The Surgical Gloves feature a polymer lining and beaded cuff to ensure easy donning and help resist tears. These Gloves have a secure fit around the wrist and hand-conforming flexibility to help prevent physical fatigue. Ideal for a wide variety of settings, including medical facilities, EMS and law enforcement, long-term care, tattoo shops, hair salons, and more. Dynarex Sterile Nitrile Surgical Gloves are high-quality, polymer-lined gloves designed for enhanced protection and performance. The Nitrile Surgical Gloves have a thickness range of 5-8 mil at the palm and fingertips to provide optimal tactile sensitivity and puncture resistance. These hand-specific, powder-free Gloves feature beaded cuffs to allow for easy donning.
Catalog No. 50-283-2029
 Restricted Item