Learn More
Med Vet International Shoulder Length Gloves .03X240X830mm 100/Box
Catalog No. 502830009
Encompass_Preferred
Top Quality. 100 per box. One size fits all. Quality 34" sanitary OB gloves are constructed of 1.25-mil clear polyethylene with sturdy seams.
Catalog No. 50-283-0009 Supplier Med Vet International Supplier No. AHSSLGSHOULDER
Top Quality. 100 per box. One size fits all. Quality 34" sanitary OB gloves are constructed of 1.25-mil clear polyethylene with sturdy seams.