Learn More
Med Vet International McKesson Perry Latex Surgical Glove, Size 5.5, Brown, 20-1355N, 400/Case
Catalog No. 503044141
Encompass_Preferred
Surgical Glove McKesson Perry Size 5.5 Sterile Latex Standard Cuff Length Smooth Brown Not Chemo Approved Size 5½ Length (average): 11 inches (295 mm) Fingertip thickness (single): 7.5 mil (0.19 mm) Cuff thickness (single): 5.5 mil (0.14 mm) Sterilization: Gamma radiation Anti-slip, smooth finish Produced on ergonomically designed formers to enhance fit and comfort Approved and accepted for embryo use Ideal for delicate procedures and ophthalmic, micro, and cardiothoracic surgery Brown color reduces glare from surgical lights
Catalog No. 50-304-4141 Supplier Med Vet International Supplier No. 1044727CS
Surgical Glove McKesson Perry Size 5.5 Sterile Latex Standard Cuff Length Smooth Brown Not Chemo Approved Size 5½ Length (average): 11 inches (295 mm) Fingertip thickness (single): 7.5 mil (0.19 mm) Cuff thickness (single): 5.5 mil (0.14 mm) Sterilization: Gamma radiation Anti-slip, smooth finish Produced on ergonomically designed formers to enhance fit and comfort Approved and accepted for embryo use Ideal for delicate procedures and ophthalmic, micro, and cardiothoracic surgery Brown color reduces glare from surgical lights