Learn More
Med Vet International McKesson Perry Performance Plus Surgical Glove, Size 6.5, Cream, 20-1065N, 400/Case
Catalog No. 503043468
Encompass_Preferred
Surgical Glove McKesson Perry Performance Plus Size 6.5 Sterile Latex Standard Cuff Length Smooth Cream Not Chemo Approved Size 6½ Cream Length (average): 11-1/2 inches Fingertip thickness (single): 9 mil (0.23 mm) Sterilization: Gamma radiation Anti-slip, smooth finish Manufactured with SureFit Technology which is designed to enhance roll down protection Continuous polymer coating for excellent dry- and damp-hand donnability
Catalog No. 50-304-3468 Supplier Med Vet International Supplier No. 1044705CS
Surgical Glove McKesson Perry Performance Plus Size 6.5 Sterile Latex Standard Cuff Length Smooth Cream Not Chemo Approved Size 6½ Cream Length (average): 11-1/2 inches Fingertip thickness (single): 9 mil (0.23 mm) Sterilization: Gamma radiation Anti-slip, smooth finish Manufactured with SureFit Technology which is designed to enhance roll down protection Continuous polymer coating for excellent dry- and damp-hand donnability