Learn More
Med Vet International Surgical Glove ENCORE Latex Micro Size 6.5 Sterile Latex Standard Cuff Length Micro-Textured Brown Chemo Tested, 5787002, 50/Box
Catalog No. 503042913
Encompass_Preferred
Like all medical gloves, Encore Latex Micro surgical gloves are powder free Smooth with micro-textured finish Size 6½ is equivalent to small: Small fits those whose palm circumference is between 6½ and 7 inches Hand specific gloves are packed sterilely in pairs
Catalog No. 50-304-2913 Supplier Med Vet International Supplier No. 362176BX
Like all medical gloves, Encore Latex Micro surgical gloves are powder free Smooth with micro-textured finish Size 6½ is equivalent to small: Small fits those whose palm circumference is between 6½ and 7 inches Hand specific gloves are packed sterilely in pairs