Learn More
Med Vet International McKesson Perry Performance Plus Surgical Glove, Size 8.5, Cream, 20-1085N, Pair
Catalog No. 503044137
Encompass_Preferred
Surgical Glove McKesson Perry Performance Plus Size 8.5 Sterile Latex Standard Cuff Length Smooth Cream Not Chemo Approved Size 8½ Cream Length (average): 11 inches (280 mm) Fingertip thickness (single): 9 mil (0.23 mm) Sterilization: Gamma radiation Anti-slip, smooth finish Manufactured with SureFit Technology which is designed to enhance roll down protection Continuous polymer coating for excellent dry- and damp-hand donnability
Catalog No. 50-304-4137 Supplier Med Vet International Supplier No. 1044709PR
Surgical Glove McKesson Perry Performance Plus Size 8.5 Sterile Latex Standard Cuff Length Smooth Cream Not Chemo Approved Size 8½ Cream Length (average): 11 inches (280 mm) Fingertip thickness (single): 9 mil (0.23 mm) Sterilization: Gamma radiation Anti-slip, smooth finish Manufactured with SureFit Technology which is designed to enhance roll down protection Continuous polymer coating for excellent dry- and damp-hand donnability