Learn More
Med Vet International McKesson Perry Performance Plus Surgical Glove, Size 9, Cream, 20-1090N, 400/Case
Catalog No. 503043466
Encompass_Preferred
Surgical Glove McKesson Perry Performance Plus Size 9 Sterile Latex Standard Cuff Length Smooth Cream Not Chemo Approved Size 9 Cream Length (average): 11 inches (280 mm) Fingertip thickness (single): 9 mil (0.23 mm) Sterilization: Gamma radiation Anti-slip, smooth finish Anti-slip, smooth finish Continuous polymer coating for excellent dry- and damp-hand donnability
Catalog No. 50-304-3466 Supplier Med Vet International Supplier No. 1044710CS
Surgical Glove McKesson Perry Performance Plus Size 9 Sterile Latex Standard Cuff Length Smooth Cream Not Chemo Approved Size 9 Cream Length (average): 11 inches (280 mm) Fingertip thickness (single): 9 mil (0.23 mm) Sterilization: Gamma radiation Anti-slip, smooth finish Anti-slip, smooth finish Continuous polymer coating for excellent dry- and damp-hand donnability