Learn More
Med Vet International McKesson Perry Performance Plus Surgical Glove, Size 7.5, Cream, 20-1075N, 200 Pairs
Catalog No. 503043472
Encompass_Preferred
These cream McKesson Perry Performance Plus latex surgical gloves help protect both patients and caregivers during critical medical procedures Continuous polymer coating for excellent dry- and damp-hand donnability Manufactured with SureFit technology, which is designed to enhance roll-down protection Size 7-1/2 gloves are 11-1/2 inches long with an extended beaded cuff length; packaged in dedicated left/right sterile pairs
Catalog No. 50-304-3472 Supplier Med Vet International Supplier No. 1044707CS
These cream McKesson Perry Performance Plus latex surgical gloves help protect both patients and caregivers during critical medical procedures Continuous polymer coating for excellent dry- and damp-hand donnability Manufactured with SureFit technology, which is designed to enhance roll-down protection Size 7-1/2 gloves are 11-1/2 inches long with an extended beaded cuff length; packaged in dedicated left/right sterile pairs