Learn More
Med Vet International McKesson Perry Synthetic Surgical Gloves Polychloroprene Surgical Glove, Size 5.5, Cream, 20-2655N, 400/Case
Catalog No. 503044183
Encompass_Preferred
Surgical Glove McKesson Perry Synthetic Surgical Gloves Size 5.5 Sterile Polychloroprene Standard Cuff Length Smooth Cream Chemo Tested Size 5½ Cream Length (average): 12.2 inches (310 mm) Fingertip thickness (single): 7.9 mil (0.20 mm) Cuff thickness (single): 6.3 mil (0.16 mm) Sterilization: Gamma radiation 100% synthetic neoprene glove that delivers the performance characteristics of natural rubber latex without the risk of natural rubber latex sensitization Anti-slip, smooth finish Manufactured with SureFit Technology which is designed to enhance roll down protection
Catalog No. 50-304-4183 Supplier Med Vet International Supplier No. 1044743CS
Surgical Glove McKesson Perry Synthetic Surgical Gloves Size 5.5 Sterile Polychloroprene Standard Cuff Length Smooth Cream Chemo Tested Size 5½ Cream Length (average): 12.2 inches (310 mm) Fingertip thickness (single): 7.9 mil (0.20 mm) Cuff thickness (single): 6.3 mil (0.16 mm) Sterilization: Gamma radiation 100% synthetic neoprene glove that delivers the performance characteristics of natural rubber latex without the risk of natural rubber latex sensitization Anti-slip, smooth finish Manufactured with SureFit Technology which is designed to enhance roll down protection