Learn More
Med Vet International Surgical Glove McKesson Perry Synthetic Surgical Gloves Size 6 Sterile Polychloroprene Standard Cuff Length Smooth Cream Chemo Tested, 20-2660N, 200/Case
Catalog No. 503043962
Encompass_Preferred
Size 6 Cream Length (average): 12.2 inches (310 mm) Fingertip thickness (single): 7.9 mil (0.20 mm) Cuff thickness (single): 6.3 mil (0.16 mm) Sterilization: Gamma radiation 100% synthetic neoprene glove that delivers the performance characteristics of natural rubber latex without the risk of natural rubber latex sensitization Anti-slip, smooth finish Manufactured with SureFit Technology which is designed to enhance roll down protection
Catalog No. 50-304-3962 Supplier Med Vet International Supplier No. 1044744CS
Size 6 Cream Length (average): 12.2 inches (310 mm) Fingertip thickness (single): 7.9 mil (0.20 mm) Cuff thickness (single): 6.3 mil (0.16 mm) Sterilization: Gamma radiation 100% synthetic neoprene glove that delivers the performance characteristics of natural rubber latex without the risk of natural rubber latex sensitization Anti-slip, smooth finish Manufactured with SureFit Technology which is designed to enhance roll down protection