Learn More
Med Vet International McKesson Perry Performance Plus Polychloroprene Surgical Glove, Size 6.5, Dark Green, 20-2565N, 50 Pairs
Catalog No. 503043493
Encompass_Preferred
Surgical Glove McKesson Perry Performance Plus Size 6.5 Sterile Polychloroprene Standard Cuff Length Smooth Dark Green Chemo Tested Size 6½ Dark Green Length (average): 12.2 inches (310 mm) Fingertip thickness (single): 7.9 mil (0.20 mm) Cuff thickness (single): 6.3 mil (0.16 mm) Sterilization: Gamma radiation 100% synthetic neoprene glove that delivers the performance characteristics of natural rubber latex without the risk of natural rubber latex sensitization Anti-slip, smooth finish Manufactured with SureFit Technology which is designed to enhance roll down protection
Catalog No. 50-304-3493 Supplier Med Vet International Supplier No. 1044737BX
Surgical Glove McKesson Perry Performance Plus Size 6.5 Sterile Polychloroprene Standard Cuff Length Smooth Dark Green Chemo Tested Size 6½ Dark Green Length (average): 12.2 inches (310 mm) Fingertip thickness (single): 7.9 mil (0.20 mm) Cuff thickness (single): 6.3 mil (0.16 mm) Sterilization: Gamma radiation 100% synthetic neoprene glove that delivers the performance characteristics of natural rubber latex without the risk of natural rubber latex sensitization Anti-slip, smooth finish Manufactured with SureFit Technology which is designed to enhance roll down protection