Learn More
Med Vet International McKesson Perry Polychloroprene Surgical Glove, Size 7.5, Cream, 20-2675N, Sterile, Pair
Catalog No. 503044165
Encompass_Preferred
100% synthetic polychloroprene glove that delivers the performance characteristics of natural rubber latex without the risk of natural rubber latex sensitization Anti-slip, smooth finish Manufactured with SureFit Technology which is designed to enhance roll down protection Size 7.5 Cream Length (average): 12.2 inches (310 mm) Fingertip thickness (single): 7.9 mil (0.20 mm) Cuff thickness (single): 6.3 mil (0.16 mm) Sterilization: Gamma radiation
Catalog No. 50-304-4165 Supplier Med Vet International Supplier No. 1044747PR
100% synthetic polychloroprene glove that delivers the performance characteristics of natural rubber latex without the risk of natural rubber latex sensitization Anti-slip, smooth finish Manufactured with SureFit Technology which is designed to enhance roll down protection Size 7.5 Cream Length (average): 12.2 inches (310 mm) Fingertip thickness (single): 7.9 mil (0.20 mm) Cuff thickness (single): 6.3 mil (0.16 mm) Sterilization: Gamma radiation