Learn More
Med Vet International McKesson Perry Synthetic Surgical Gloves Polychloroprene Surgical Glove, Size 9, Cream, 20-2690N, 400/Case
Catalog No. 503043991
Encompass_Preferred
Surgical Glove McKesson Perry Synthetic Surgical Gloves Size 9 Sterile Polychloroprene Standard Cuff Length Smooth Cream Chemo Tested Size 9 Cream Length (average): 12.2 inches (310 mm) Fingertip thickness (single): 7.9 mil (0.20 mm) Cuff thickness (single): 6.3 mil (0.16 mm) Sterilization: Gamma radiation 100% synthetic neoprene glove that delivers the performance characteristics of natural rubber latex without the risk of natural rubber latex sensitization Manufactured with SureFit Technology which is designed to enhance roll down protection Manufactured with SureFit Technology which is designed to enhance roll down protection
Catalog No. 50-304-3991 Supplier Med Vet International Supplier No. 1044750CS
Surgical Glove McKesson Perry Synthetic Surgical Gloves Size 9 Sterile Polychloroprene Standard Cuff Length Smooth Cream Chemo Tested Size 9 Cream Length (average): 12.2 inches (310 mm) Fingertip thickness (single): 7.9 mil (0.20 mm) Cuff thickness (single): 6.3 mil (0.16 mm) Sterilization: Gamma radiation 100% synthetic neoprene glove that delivers the performance characteristics of natural rubber latex without the risk of natural rubber latex sensitization Manufactured with SureFit Technology which is designed to enhance roll down protection Manufactured with SureFit Technology which is designed to enhance roll down protection