Learn More
Med Vet International Surgical Glove GAMMEX Non-Latex Sensitive Size 6.5 Sterile Polychloroprene Standard Cuff Length Micro-Textured Cream Chemo Tested, 20277265, 200/Case
Catalog No. 503044246
Encompass_Preferred
The micro-textured finish of Gammex Non-Latex Sensitive sterile gloves enable secure handling of surgical instruments – a key factor when considering which surgical gloves to buy Easy donning and doffing: Internal coating of Gammex Non-Latex Sensitive sterile gloves facilitate both damp and dry hand donning Tested to ASTM D6978, these Gammex nonlatex gloves are safe to use with a wide range of chemotherapy drugs and provide resistance to alcohol-based hand disinfectants Size 6-1/2 is between an extra small and small size fits those whose palm circumference is between 6 and 6-1/2 inches Hand specific gloves are packaged in sterile pairs
Catalog No. 50-304-4246 Supplier Med Vet International Supplier No. 833673CS
The micro-textured finish of Gammex Non-Latex Sensitive sterile gloves enable secure handling of surgical instruments – a key factor when considering which surgical gloves to buy Easy donning and doffing: Internal coating of Gammex Non-Latex Sensitive sterile gloves facilitate both damp and dry hand donning Tested to ASTM D6978, these Gammex nonlatex gloves are safe to use with a wide range of chemotherapy drugs and provide resistance to alcohol-based hand disinfectants Size 6-1/2 is between an extra small and small size fits those whose palm circumference is between 6 and 6-1/2 inches Hand specific gloves are packaged in sterile pairs