Learn More
Med Vet International Surgical Glove GAMMEX Non-Latex Sensitive Size 7.5 Sterile Polychloroprene Standard Cuff Length Micro-Textured Cream Chemo Tested, 20277275, 50/Box
Catalog No. 503044247
Encompass_Preferred
The micro-textured finish of Gammex Non-Latex Sensitive sterile gloves enable secure handling of surgical instruments – a key factor when considering which surgical gloves to buy Easy donning and doffing: Internal coating of Gammex Non-Latex Sensitive sterile gloves facilitate both damp and dry hand donning Tested to ASTM D6978, these Gammex nonlatex gloves are safe to use with a wide range of chemotherapy drugs and provide resistance to alcohol-based hand disinfectants Size 7-1/2 is between small and medium size: This size is for those who have a palm circumference between 7 and 7½ inches Hand specific gloves are packaged in sterile pairs
Catalog No. 50-304-4247 Supplier Med Vet International Supplier No. 833675BX
The micro-textured finish of Gammex Non-Latex Sensitive sterile gloves enable secure handling of surgical instruments – a key factor when considering which surgical gloves to buy Easy donning and doffing: Internal coating of Gammex Non-Latex Sensitive sterile gloves facilitate both damp and dry hand donning Tested to ASTM D6978, these Gammex nonlatex gloves are safe to use with a wide range of chemotherapy drugs and provide resistance to alcohol-based hand disinfectants Size 7-1/2 is between small and medium size: This size is for those who have a palm circumference between 7 and 7½ inches Hand specific gloves are packaged in sterile pairs