Learn More
Med Vet International Surgical Glove GAMMEX Non-Latex PI Green Size 6.5 Sterile Polyisoprene Standard Cuff Length Micro-Textured Light Green Chemo Tested, 20685265, 200/Case
Catalog No. 503043637
Encompass_Preferred
Soft yet strong formulation ensures wearers benefit from exceptional fit and feel Smooth surface with a micro-textured finish offers the wearer secure handling and instrument grip during the most rigorous procedures Optimal allergy protection Gloves' internal coating facilitates both dry and damp hand donning, either when first donned or during intra-operative changes Tested to ASTM D6978, these Gammex gloves are safe to use with a wide range of chemotherapy drugs Green polyisoprene sterile surgical gloves are size 6-1/2 and have beaded cuffs
Catalog No. 50-304-3637 Supplier Med Vet International Supplier No. 648142CS
Soft yet strong formulation ensures wearers benefit from exceptional fit and feel Smooth surface with a micro-textured finish offers the wearer secure handling and instrument grip during the most rigorous procedures Optimal allergy protection Gloves' internal coating facilitates both dry and damp hand donning, either when first donned or during intra-operative changes Tested to ASTM D6978, these Gammex gloves are safe to use with a wide range of chemotherapy drugs Green polyisoprene sterile surgical gloves are size 6-1/2 and have beaded cuffs