Learn More
Med Vet International Surgical Glove GAMMEX Non-Latex PI Size 6.5 Sterile Polyisoprene Standard Cuff Length Micro-Textured White Chemo Tested, 20685765, 200/Case
Catalog No. 503043596
Encompass_Preferred
Smooth with micro-textured finish for exceptional instrument handling and grip Free from chemical accelerators known to cause chemical Type IV allergies and sensitivities for optimized allergenic profile Dermashield Technology internal coating facilitates dry and damp donning as well as intra-operative changes Like all medical gloves, these are powder free Beaded cuffs have Surefit technology Gloves are packed in sterile pairs Size 6-1/2 is between an extra small and small size and fits those whose palm circumference is between 6 and 6-1/2 inches
Catalog No. 50-304-3596 Supplier Med Vet International Supplier No. 765477CS
Smooth with micro-textured finish for exceptional instrument handling and grip Free from chemical accelerators known to cause chemical Type IV allergies and sensitivities for optimized allergenic profile Dermashield Technology internal coating facilitates dry and damp donning as well as intra-operative changes Like all medical gloves, these are powder free Beaded cuffs have Surefit technology Gloves are packed in sterile pairs Size 6-1/2 is between an extra small and small size and fits those whose palm circumference is between 6 and 6-1/2 inches