Learn More
Med Vet International Surgical Glove GAMMEX Non-Latex PI Size 7 Sterile Polyisoprene Standard Cuff Length Micro-Textured White Chemo Tested, 20685770, 50/Box
Catalog No. 503044326
Encompass_Preferred
Smooth with micro-textured finish for exceptional instrument handling and grip Free from chemical accelerators known to cause chemical Type IV allergies and sensitivities for optimized allergenic profile Dermashield Technology internal coating facilitates dry and damp donning as well as intra-operative changes Like all medical gloves, these are powder free Beaded cuffs have Surefit technology Gloves are packed in sterile pairs Size 7 is equivalent to size small: Small fits those palm circumference is between 6-1/2 and 7 inches
Catalog No. 50-304-4326 Supplier Med Vet International Supplier No. 765478BX
Smooth with micro-textured finish for exceptional instrument handling and grip Free from chemical accelerators known to cause chemical Type IV allergies and sensitivities for optimized allergenic profile Dermashield Technology internal coating facilitates dry and damp donning as well as intra-operative changes Like all medical gloves, these are powder free Beaded cuffs have Surefit technology Gloves are packed in sterile pairs Size 7 is equivalent to size small: Small fits those palm circumference is between 6-1/2 and 7 inches