Learn More
Med Vet International Surgical Glove GAMMEX Non-Latex PI Green Size 7.5 Sterile Polyisoprene Standard Cuff Length Micro-Textured Light Green Chemo Tested, 20685275, 200/Case
Catalog No. 503044192
Encompass_Preferred
Polymer coated with Dermashield technology Smooth with micro-textured finish Standard thickness Moderate grip level 7½ size is equivalent to small/medium size: They will fit those whose palm circumference is between 7½ and 8 inches Green
Catalog No. 50-304-4192 Supplier Med Vet International Supplier No. 648144CS
Polymer coated with Dermashield technology Smooth with micro-textured finish Standard thickness Moderate grip level 7½ size is equivalent to small/medium size: They will fit those whose palm circumference is between 7½ and 8 inches Green