Learn More
Med Vet International McKesson Perry Performance Plus Polyisoprene Surgical Glove, Size 7.5, Green, 20-2075N, Pair
Catalog No. 503044161
Encompass_Preferred
Surgical Glove McKesson Perry Performance Plus Size 7.5 Sterile Polyisoprene Standard Cuff Length Smooth Green Chemo Tested Size 7½ Green Length (average): 12.2 inches (310 mm) Fingertip thickness (single): 9 mil (0.23 mm) Cuff thickness (single): 7.1 mil (0.18 mm) Sterilization: Gamma radiation 100% synthetic polyisoprene glove delivers the performance characteristics of natural rubber latex without the risk of natural rubber latex sensitization Anti-slip, smooth finish Manufactured with SureFit Technology which is designed to enhance roll down protection
Catalog No. 50-304-4161 Supplier Med Vet International Supplier No. 1044723PR
Surgical Glove McKesson Perry Performance Plus Size 7.5 Sterile Polyisoprene Standard Cuff Length Smooth Green Chemo Tested Size 7½ Green Length (average): 12.2 inches (310 mm) Fingertip thickness (single): 9 mil (0.23 mm) Cuff thickness (single): 7.1 mil (0.18 mm) Sterilization: Gamma radiation 100% synthetic polyisoprene glove delivers the performance characteristics of natural rubber latex without the risk of natural rubber latex sensitization Anti-slip, smooth finish Manufactured with SureFit Technology which is designed to enhance roll down protection