Learn More
Med Vet International Surgical Glove Protexis™ PI Size 7.5 Sterile Polyisoprene Standard Cuff Length Smooth Ivory Chemo Tested, 2D72PT75X, Pair
Catalog No. 503043319
Encompass_Preferred
Engineered to protect in a wide array of clinical cases Proprietary hand mold with an independent thumb design allows for an anatomical fit and natural movement in the fingers, thumb and palm Size 7½
Catalog No. 50-304-3319 Supplier Med Vet International Supplier No. 806827PR
Engineered to protect in a wide array of clinical cases Proprietary hand mold with an independent thumb design allows for an anatomical fit and natural movement in the fingers, thumb and palm Size 7½