Learn More
Med Vet International McKesson Perry Polyisoprene Standard Cuff Length Surgical Glove, Size 8½, Green, 20-2085N, Pair
Catalog No. 503043911
Encompass_Preferred
Surgical Glove McKesson Perry Performance Plus Size 8.5 Sterile Pair Polyisoprene Extended Cuff Length Smooth Green Chemo Tested Size 8½ Green Length: 12? inches Fingertip thickness (single): 9 mil (0.23 mm) Cuff thickness (single): 7.1 mil (0.18 mm) Sterilization: Gamma radiation 100% synthetic polyisoprene glove that delivers the performance characteristics of natural rubber latex without the risk of natural rubber latex sensitization Anti-slip, smooth finish Manufactured with SureFit™ Technology which is designed to enhance roll down protection
Catalog No. 50-304-3911 Supplier Med Vet International Supplier No. 1044725PR
Surgical Glove McKesson Perry Performance Plus Size 8.5 Sterile Pair Polyisoprene Extended Cuff Length Smooth Green Chemo Tested Size 8½ Green Length: 12? inches Fingertip thickness (single): 9 mil (0.23 mm) Cuff thickness (single): 7.1 mil (0.18 mm) Sterilization: Gamma radiation 100% synthetic polyisoprene glove that delivers the performance characteristics of natural rubber latex without the risk of natural rubber latex sensitization Anti-slip, smooth finish Manufactured with SureFit™ Technology which is designed to enhance roll down protection