Learn More
Med Vet International McKesson Perry Performance Plus Polyisoprene Surgical Glove, Size 9, Green, 20-2090N, 400/Case
Catalog No. 503043330
Encompass_Preferred
Surgical Glove McKesson Perry Performance Plus Size 9 Sterile Polyisoprene Standard Cuff Length Smooth Green Chemo Tested Size 9 Green Length: 12? inches Fingertip thickness (single): 9 mil (0.23 mm) Cuff thickness (single): 7.1 mil (0.18 mm) Sterilization: Gamma radiation 100% synthetic polyisoprene glove delivers the performance characteristics of natural rubber latex without the risk of natural rubber latex sensitization Approved and accepted for embryo use Ideal for delicate procedures and ophthalmic, micro, and cardiothoracic surgery Manufactured with SureFit Technology which is designed to enhance roll down protection
Catalog No. 50-304-3330 Supplier Med Vet International Supplier No. 1044726CS
Surgical Glove McKesson Perry Performance Plus Size 9 Sterile Polyisoprene Standard Cuff Length Smooth Green Chemo Tested Size 9 Green Length: 12? inches Fingertip thickness (single): 9 mil (0.23 mm) Cuff thickness (single): 7.1 mil (0.18 mm) Sterilization: Gamma radiation 100% synthetic polyisoprene glove delivers the performance characteristics of natural rubber latex without the risk of natural rubber latex sensitization Approved and accepted for embryo use Ideal for delicate procedures and ophthalmic, micro, and cardiothoracic surgery Manufactured with SureFit Technology which is designed to enhance roll down protection