Learn More
Med Vet International Sensicare Surgical Gloves With Aloe, Latex Free, Size 8, 100 Pair/Case
Catalog No. 502831217
Encompass_Preferred
SensiCare with Aloe were the world's first synthetic polyisoprene surgical gloves on the market, Anti-roll-down cuff technology significantly reduces the roll down common with many surgical gloves, The anatomically curved style offers comfort, Gloves contain freeze-dried aloe
Catalog No. 50-283-1217 Supplier Med Vet International Supplier No. MSG1080
SensiCare with Aloe were the world's first synthetic polyisoprene surgical gloves on the market, Anti-roll-down cuff technology significantly reduces the roll down common with many surgical gloves, The anatomically curved style offers comfort, Gloves contain freeze-dried aloe