Learn More
M BRAUN M BRAUN Anatomical Glove Box Gloves, 0.4 mm Thickness, Size 9.75, 1 Pair
Catalog No. NC1920038
Encompass
The M BRAUN gloves are designed for precise handling in controlled environments, featuring a size 9.75 fit and anatomical design for comfort. With a 0.4 mm thickness and 220 mm port diameter, these gloves ensure secure integration and dexterity for scientific applications. Their left/right specificity sets them apart, enhancing user experience during use.
Catalog No. NC1920038 Supplier M BRAUN Supplier No. 2800027
The M BRAUN gloves are designed for precise handling in controlled environments, featuring a size 9.75 fit and anatomical design for comfort. With a 0.4 mm thickness and 220 mm port diameter, these gloves ensure secure integration and dexterity for scientific applications. Their left/right specificity sets them apart, enhancing user experience during use.