Learn More
Warwick Mills Inc GLOVE TURTLE SKIN UTLITY MED
SDP
Supplier:  Warwick Mills Inc UPW4D1M
Encompass_Preferred
Turtleskin Utility Gloves, Medium, Features the highest resistance to very fine penetrators and softens the more you wear it, Puncture protection: Level 3 Cut protection: Excellent
Catalog No. NC9269325