Learn More
Med Vet International Dynarex Next Generation Stretch Vinyl Exam Gloves, Large, Powder Free, 1000/Case
Catalog No. 502831396
Encompass_Preferred
Dynarex Next Generation Stretch Vinyl Exam Gloves are premium-quality, powder-free vinyl gloves designed for improved protection and flexibility. The Exam Gloves are crafted from a soft vinyl resin for added elasticity and increased sensitivity. These ambidextrous Gloves feature a minimum thickness of 3 mil. Dynarex Next Generation Stretch Vinyl Exam Gloves are designed with a polymer lining to help resist tears and beaded cuffs to allow for easy donning. These Gloves have a secure fit around the wrist and hand-conforming flexibility to help prevent physical fatigue. Ideal for a variety of settings, including medical facilities, long-term care facilities, food service, and hair salons. Dynarex Next Generation Stretch Vinyl Exam Gloves are high-quality, polymer-lined gloves designed for enhanced protection and performance. These ambidextrous, powder-free Gloves are crafted from soft vinyl for added flexibility and increased tactile sensitivity. 
Catalog No. 50-283-1396 Supplier Med Vet International Supplier No. 6824
Dynarex Next Generation Stretch Vinyl Exam Gloves are premium-quality, powder-free vinyl gloves designed for improved protection and flexibility. The Exam Gloves are crafted from a soft vinyl resin for added elasticity and increased sensitivity. These ambidextrous Gloves feature a minimum thickness of 3 mil. Dynarex Next Generation Stretch Vinyl Exam Gloves are designed with a polymer lining to help resist tears and beaded cuffs to allow for easy donning. These Gloves have a secure fit around the wrist and hand-conforming flexibility to help prevent physical fatigue. Ideal for a variety of settings, including medical facilities, long-term care facilities, food service, and hair salons. Dynarex Next Generation Stretch Vinyl Exam Gloves are high-quality, polymer-lined gloves designed for enhanced protection and performance. These ambidextrous, powder-free Gloves are crafted from soft vinyl for added flexibility and increased tactile sensitivity. 