Learn More
Med Vet International Dynarex Safe-Touch Vinyl Exam Gloves, Large, Powder-Free, Clear, 1000/Case
Catalog No. 502831777
Encompass_Preferred
Dynarex Safe-Touch Vinyl Exam Gloves are premium-quality powder-free vinyl gloves designed for improved protection and flexibility. The Exam Gloves are crafted from a soft vinyl resin for added elasticity and increased sensitivity. These ambidextrous Gloves feature a minimum thickness of 3 mil. Dynarex Safe-Touch Vinyl Exam Gloves are designed with a polymer lining to help resist tears and beaded cuffs to allow for easy donning. These Gloves have a secure fit around the wrist and hand-conforming flexibility to help prevent physical fatigue. Ideal for a variety of settings including medical facilities long-term care facilities food service and hair salons. Dynarex Safe-Touch Vinyl Exam Gloves are high-quality polymer-lined gloves designed for enhanced protection and performance. These ambidextrous powder-free Gloves are crafted from soft vinyl for added flexibility and increased tactile sensitivity. Ideal for a wide range of healthcare professional and food service environments.
Catalog No. 50-283-1777 Supplier Med Vet International Supplier No. DYN2613
Dynarex Safe-Touch Vinyl Exam Gloves are premium-quality powder-free vinyl gloves designed for improved protection and flexibility. The Exam Gloves are crafted from a soft vinyl resin for added elasticity and increased sensitivity. These ambidextrous Gloves feature a minimum thickness of 3 mil. Dynarex Safe-Touch Vinyl Exam Gloves are designed with a polymer lining to help resist tears and beaded cuffs to allow for easy donning. These Gloves have a secure fit around the wrist and hand-conforming flexibility to help prevent physical fatigue. Ideal for a variety of settings including medical facilities long-term care facilities food service and hair salons. Dynarex Safe-Touch Vinyl Exam Gloves are high-quality polymer-lined gloves designed for enhanced protection and performance. These ambidextrous powder-free Gloves are crafted from soft vinyl for added flexibility and increased tactile sensitivity. Ideal for a wide range of healthcare professional and food service environments.