Learn More
Med Vet International Dynarex Vinyl Exam Gloves in A Bag, Large, Powder-Free, Clear, 2 Pairs/Bag, 500 Bags/Case
Catalog No. 502829254
Encompass_Preferred
Dynarex DynaSafety Vinyl Gloves in a Bag are premium-quality, powder-free vinyl gloves designed for improved protection and flexibility. The Gloves are crafted from a soft vinyl resin for added elasticity and increased sensitivity. These ambidextrous Gloves feature a minimum thickness of 3 mils at the palm and 2 mils at the fingertips. Dynarex Vinyl Gloves in a Bag are designed with beaded cuffs to allow for easy donning. These Gloves have a secure fit around the wrist and hand-conforming flexibility to help prevent physical fatigue. Ideal for a variety of settings, including first-aid, medical facilities, long-term care, food service, and hair salons. Dynarex DynaSafety Vinyl Gloves in a Bag are high-quality, polymer-lined gloves designed for enhanced protection and performance. These ambidextrous, powder-free Gloves are crafted from soft vinyl for added flexibility and increased tactile sensitivity. Ideal for a wide range of healthcare, professional, and food service environments.
Catalog No. 50-282-9254 Supplier Med Vet International Supplier No. 2617
Dynarex DynaSafety Vinyl Gloves in a Bag are premium-quality, powder-free vinyl gloves designed for improved protection and flexibility. The Gloves are crafted from a soft vinyl resin for added elasticity and increased sensitivity. These ambidextrous Gloves feature a minimum thickness of 3 mils at the palm and 2 mils at the fingertips. Dynarex Vinyl Gloves in a Bag are designed with beaded cuffs to allow for easy donning. These Gloves have a secure fit around the wrist and hand-conforming flexibility to help prevent physical fatigue. Ideal for a variety of settings, including first-aid, medical facilities, long-term care, food service, and hair salons. Dynarex DynaSafety Vinyl Gloves in a Bag are high-quality, polymer-lined gloves designed for enhanced protection and performance. These ambidextrous, powder-free Gloves are crafted from soft vinyl for added flexibility and increased tactile sensitivity. Ideal for a wide range of healthcare, professional, and food service environments.