Learn More
Med Vet International Vet Mitt X-Ray Gloves, J0677M, Each
Catalog No. 5000495357
Encompass_Preferred
Features a palm slit allowing fingers to exit for freedom of movement but still cover top of hand during radiology. Royal blue nylon. Standard protection: 0.5mm Pb equivalency.•The protective gloves and mitten are mainly used by the operators to guard against the rays during their operation.•The medical Xray protective glove and mitten are processed by rubber.
Catalog No. 50-004-95357 Supplier Med Vet International Supplier No. JOR677M
Features a palm slit allowing fingers to exit for freedom of movement but still cover top of hand during radiology. Royal blue nylon. Standard protection: 0.5mm Pb equivalency.•The protective gloves and mitten are mainly used by the operators to guard against the rays during their operation.•The medical Xray protective glove and mitten are processed by rubber.