Learn More
Glas Col LLC Glove Bag
SDP
Supplier:  Glas Col LLC 108D X1717HG
Encompass_Preferred
Glove bag, 17" x 17" x 11", one 11 1/2" equipment opening in the front between two integral turned in gloves.Made of 3mil low density polyethylene. HandyGlide closure.
Catalog No. NC0913747