Learn More
TA Instruments Glovebox Adaptor Kit for Discovery TGA 55 550 5500, Gas Tight Feedthroughs, KF-40 Openings, Maintain Stability, Requires UPS
Catalog No. 5000580640 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments Glovebox Adaptor Kit allows installation and operation of Discovery TGA 55, 550, and 5500 in a glovebox. It provides gas tight feedthroughs for cabling, gas, and cooling using three KF-40 openings. To maintain stability, the glovebox pump should not run during measurements. Requires UL1778 or CE approved UPS.
Catalog No. 50-005-80640 Supplier TA Instruments Supplier No. 534910.901
Waters TA Instruments Glovebox Adaptor Kit allows installation and operation of Discovery TGA 55, 550, and 5500 in a glovebox. It provides gas tight feedthroughs for cabling, gas, and cooling using three KF-40 openings. To maintain stability, the glovebox pump should not run during measurements. Requires UL1778 or CE approved UPS.