Learn More
Med Vet International Curad PF Nitrile Exam Gloves, Blue, Large, 1500/Case
Catalog No. 502232656
Encompass_Preferred
This nitrile glove is an excellent latex-free alternative. It is lightly textured for better grip and blue in color for greater visibility. Beaded cuff facilitates donning. These gloves are tested for use with chemotherapy drugs. Large.150/Box, 10 Box/Case
This item is not returnable. View return policy
Catalog No. 50-223-2656 Supplier Med Vet International Supplier No. CUR9316
This nitrile glove is an excellent latex-free alternative. It is lightly textured for better grip and blue in color for greater visibility. Beaded cuff facilitates donning. These gloves are tested for use with chemotherapy drugs. Large.150/Box, 10 Box/Case