Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gloveworks Black Nitrile Industrial Grade Gloves
GREENER_CHOICE

Catalog No. 19227019
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-227-019 2X-Large
19-227-015 Small
19-227-016 Medium
19-227-017 Large
19-227-018 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-227-019 Supplier Gloveworks Supplier No. GPNB49100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Black nitrile industrial gloves offer the puncture resistance of latex, and provide protection against common and specialty chemicals. Micro-roughened surface and textured fingertips enhance grip, while the polymer coated interior leads to easier glove changes.

  • Constructed from medium-weight premium nitrile
  • At 5 mils thick, they are more than 50% thicker than standard nitrile gloves
  • Nitrile has three times the puncture resistance of latex
  • Strong barrier protection against common and specialty chemicals, including cleaning solutions
  • Micro-roughened surface and textured fingertips enhance grip
  • Polymer coated on the inside for easier glove changes
  • Black industrial gloves look professional while concealing dirt and grime
  • Ideal for professionals in automotive, industrial, janitorial, sanitation, food service, and safety applications
  • Latex-free for those allergic to natural rubber

Specifications

Applications Industrial
Color Black
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 6 mil
Finger Thickness (Metric) 0.15 mm
Laboratory/Cleanroom Use No
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5 mil
Palm Thickness (Metric) 0.13 mm
Size 2X-Large
Series GPNB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.